Assay Details
Gene Symbol: ENSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151662.2 | XP_015007148.2 | 380 | 102 | ||
XM_015151671.2 | XP_015007157.2 | 380 | 102 | |||
XM_015151678.2 | XP_015007164.1 | 343 | 102 | |||
XM_028829314.1 | XP_028685147.1 | 170 | 102 | |||
GenBank mRNA | JU319729.1 | - | 265 | 102 | ||
JU319730.1 | - | 265 | 102 | |||
JU471596.1 | - | 315 | 102 | |||
JU471597.1 | - | 315 | 102 | |||
JU471598.1 | - | 315 | 102 | |||
JV044628.1 | - | 318 | 102 | |||
JV044629.1 | - | 318 | 102 | |||
JV489128.1 | - | 333 | 102 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
