Assay Details
Gene Symbol: LOC713572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028851424.1 | XP_028707257.1 | 2213 | 117 | ||
XM_028851426.1 | XP_028707259.1 | 319 | 117 | |||
GenBank mRNA | JV564947.1 | - | 4 - 5 | 1114 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
