Assay Details
Gene Symbol: VGLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104200.4 | XP_001104200.1 | 694 | 57 | ||
XM_015129166.2 | XP_014984652.1 | 698 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
