Assay Details
Gene Symbol: TENT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266603.1 | NP_001253532.1 | 122 | 84 | ||
GenBank mRNA | JU476132.1 | - | 96 | 84 | ||
JV043890.1 | - | 95 | 84 | |||
JV043891.1 | - | 95 | 84 | |||
JV635594.1 | - | 122 | 84 | |||
JV637740.1 | - | 122 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266603.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
