Assay Details
Gene Symbol: TNFAIP8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257419.1 | NP_001244348.1 | 74 | 123 | ||
GenBank mRNA | JU322215.1 | - | 74 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257419.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
