Assay Details
Gene Symbol: C1QTNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114277.2 | XP_014969763.2 | 1048 | 141 | ||
GenBank mRNA | JV706327.1 | - | 2 | 304 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
