Assay Details
Gene Symbol: MPV17L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194635.1 | NP_001181564.1 | 658 | 80 | ||
GenBank mRNA | JU327145.1 | - | 559 | 80 | ||
JU495055.1 | - | 542 | 80 | |||
JV048477.1 | - | 597 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
