Assay Details
Gene Symbol: NT5C3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119667.2 | XP_014975153.1 | 875 | 90 | ||
XM_015119669.2 | XP_014975155.1 | 785 | 90 | |||
XM_015119670.2 | XP_014975156.1 | 770 | 90 | |||
XM_028836484.1 | XP_028692317.1 | 957 | 90 | |||
GenBank mRNA | JU322519.1 | - | 8 - 9 | 806 | 90 | |
JU474225.1 | - | 8 - 9 | 806 | 90 | ||
JV046119.1 | - | 8 - 9 | 770 | 90 | ||
JV493863.1 | - | 7 - 8 | 751 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
