Assay Details
Gene Symbol: TSSK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104572.4 | XP_001104572.1 | 672 | 87 | ||
GenBank mRNA | JU324636.1 | - | 1 | 61 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
