Assay Details
Gene Symbol: BORCS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194906.2 | NP_001181835.1 | 316 | 131 | ||
XM_028853193.1 | XP_028709026.1 | 296 | 131 | |||
GenBank mRNA | JU321683.1 | - | 276 | 131 | ||
JU474261.1 | - | 274 | 131 | |||
JV045399.1 | - | 276 | 131 | |||
JV669687.1 | - | 300 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() |
