Assay Details
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113826.2 | XP_014969312.2 | 624 | 109 | ||
XM_028826818.1 | XP_028682651.1 | 602 | 109 | |||
GenBank mRNA | JU338741.1 | - | 1 - 2 | 451 | 109 | |
JV706851.1 | - | 1 - 2 | 674 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
