Assay Details
Gene Symbol: P2RY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028833806.1 | XP_028689639.1 | 2835 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028833806.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
