Assay Details
Gene Symbol: LOC114674356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840868.1 | XP_028696701.1 | 161 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028840868.1 transcripts.
Set Membership: |
![]() ![]() |
