Assay Details
Gene Symbol: BLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097543.4 | XP_001097543.2 | 962 | 162 | ||
XM_015143029.2 | XP_014998515.2 | 962 | 162 | |||
XM_028852012.1 | XP_028707845.1 | 1017 | 162 | |||
GenBank mRNA | JU476758.1 | - | 3 - 4 | 881 | 162 |

More Information
Set Membership: |
![]() ![]() ![]() |
