Assay Details
Gene Symbol: DDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260837.1 | NP_001247766.1 | 9 - 10 | 1413 | 111 | |
GenBank mRNA | JU472367.1 | - | 9 - 10 | 1413 | 111 |
Gene Symbol: LOC114672263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028833469.1 | XP_028689302.1 | 1002 | 111 |
Gene Symbol: LOC114672264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU327791.1 | - | 3 - 4 | 226 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
