Assay Details
Gene Symbol: PMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110539.2 | XP_014966025.2 | 2854 | 99 | ||
XM_015110540.2 | XP_014966026.2 | 2980 | 99 | |||
XM_015110543.2 | XP_014966029.2 | 2737 | 99 | |||
XM_015110544.2 | XP_014966030.2 | 2682 | 99 | |||
XM_028830931.1 | XP_028686764.1 | 2682 | 99 | |||
XM_028830932.1 | XP_028686765.1 | 2682 | 99 | |||
XM_028830933.1 | XP_028686766.1 | 2682 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
