Assay Details
Gene Symbol: IGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110935.2 | XP_014966421.2 | 556 | 102 | ||
GenBank mRNA | JU327317.1 | - | 268 | 102 | ||
JV060016.1 | - | 391 | 102 | |||
JV650362.1 | - | 392 | 102 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
