Assay Details
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265649.2 | NP_001252578.1 | 4 - 5 | 401 | 93 | |
XM_015109913.2 | XP_014965399.1 | 1152 | 93 | |||
XM_015109918.2 | XP_014965404.1 | 1088 | 93 | |||
GenBank mRNA | JV564974.1 | - | 4 - 5 | 406 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
