Assay Details
Gene Symbol: GNRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195436.2 | NP_001182365.1 | 2 - 3 | 240 | 76 | |
GenBank mRNA | JU323469.1 | - | 2 - 3 | 490 | 76 | |
JU472587.1 | - | 2 - 3 | 1204 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
