Assay Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089664.4 | XP_001089664.3 | 3557 | 133 | ||
GenBank mRNA | JU493968.1 | - | 1 - 2 | 848 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() |
