Assay Details
Gene Symbol: TAFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108739.4 | XP_001108739.1 | 1311 | 138 | ||
XM_015148485.2 | XP_015003971.1 | 407 | 138 | |||
XM_015148495.2 | XP_015003981.1 | 411 | 138 | |||
XM_028830327.1 | XP_028686160.1 | 1087 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001108739.4, XM_015148485.2, XM_015148495.2, XM_028830327.1 transcripts.
Set Membership: |
![]() ![]() |
