Assay Details
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103280.4 | XP_001103280.2 | 399 | 90 | ||
XM_015129853.2 | XP_014985339.2 | 405 | 90 | |||
GenBank mRNA | JU475040.1 | - | 349 | 90 | ||
JU475041.1 | - | 355 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
