Assay Details
Gene Symbol: NAA30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266880.2 | NP_001253809.1 | 868 | 103 | ||
GenBank mRNA | JU326896.1 | - | 577 | 103 | ||
JV046166.1 | - | 868 | 103 | |||
JV641940.1 | - | 903 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
