Assay Details
Gene Symbol: TNFAIP8L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193689.2 | NP_001180618.1 | 316 | 159 | ||
XM_015142194.2 | XP_014997680.1 | 350 | 159 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015142194.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
