Assay Details
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115393.2 | XP_014970879.1 | 898 | 146 | ||
XM_015115395.2 | XP_014970881.1 | 898 | 146 | |||
XM_028833900.1 | XP_028689733.1 | 540 | 146 | |||
GenBank mRNA | JU341133.1 | - | 6 - 7 | 960 | 146 | |
JU341134.1 | - | 6 - 7 | 960 | 146 | ||
JV565045.1 | - | 5 - 6 | 798 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() |
