Assay Details
Gene Symbol: RTKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261179.1 | NP_001248108.1 | 4 - 5 | 525 | 118 | |
XM_015112538.2 | XP_014968024.1 | 2019 | 118 | |||
XM_015112539.2 | XP_014968025.1 | 1459 | 118 | |||
XM_015112540.2 | XP_014968026.1 | 1052 | 118 | |||
XM_015112541.2 | XP_014968027.1 | 1597 | 118 | |||
XM_015112543.2 | XP_014968029.1 | 1055 | 118 | |||
XM_028831552.1 | XP_028687385.1 | 1049 | 118 | |||
XM_028831553.1 | XP_028687386.1 | 3831 | 118 | |||
XM_028831554.1 | XP_028687387.1 | 2025 | 118 | |||
XM_028831555.1 | XP_028687388.1 | 934 | 118 | |||
XM_028831556.1 | XP_028687389.1 | 1050 | 118 | |||
XM_028831557.1 | XP_028687390.1 | 2019 | 118 | |||
XM_028831558.1 | XP_028687391.1 | 977 | 118 | |||
XM_028831559.1 | XP_028687392.1 | 2030 | 118 | |||
XM_028831560.1 | XP_028687393.1 | 1053 | 118 | |||
GenBank mRNA | JU321015.1 | - | 5 - 6 | 954 | 118 | |
JU322391.1 | - | 4 - 5 | 525 | 118 | ||
JV043526.1 | - | 5 - 6 | 896 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
