Assay Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028850979.1 | XP_028706812.1 | 1322 | 132 | ||
GenBank mRNA | JU321531.1 | - | 1305 | 132 | ||
JV704273.1 | - | 1312 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
