Assay Details
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257533.1 | NP_001244462.1 | 384 | 143 | ||
XM_015133538.2 | XP_014989024.1 | 446 | 143 | |||
XM_015133541.2 | XP_014989027.1 | 442 | 143 | |||
XM_015133542.2 | XP_014989028.1 | 275 | 143 | |||
XM_015133545.2 | XP_014989031.1 | 271 | 143 | |||
XM_015133547.2 | XP_014989033.1 | 445 | 143 | |||
XM_028844914.1 | XP_028700747.1 | 599 | 143 | |||
XM_028844916.1 | XP_028700749.1 | 597 | 143 | |||
XM_028844918.1 | XP_028700751.1 | 425 | 143 | |||
XM_028844921.1 | XP_028700754.1 | 421 | 143 | |||
XM_028844922.1 | XP_028700755.1 | 599 | 143 | |||
GenBank mRNA | JU336612.1 | - | 178 | 143 | ||
JU336613.1 | - | 352 | 143 | |||
JU336614.1 | - | 384 | 143 | |||
JU477425.1 | - | 414 | 143 | |||
JV044725.1 | - | 392 | 143 | |||
JV048386.1 | - | 404 | 143 | |||
JV630355.1 | - | 287 | 143 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
