Assay Details
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257533.1 | NP_001244462.1 | 905 | 142 | ||
XM_015133538.2 | XP_014989024.1 | 1090 | 142 | |||
XM_015133541.2 | XP_014989027.1 | 963 | 142 | |||
XM_015133542.2 | XP_014989028.1 | 919 | 142 | |||
XM_015133545.2 | XP_014989031.1 | 792 | 142 | |||
XM_015133547.2 | XP_014989033.1 | 693 | 142 | |||
XM_028844914.1 | XP_028700747.1 | 1243 | 142 | |||
XM_028844915.1 | XP_028700748.1 | 1160 | 142 | |||
XM_028844916.1 | XP_028700749.1 | 1118 | 142 | |||
XM_028844917.1 | XP_028700750.1 | 1006 | 142 | |||
XM_028844918.1 | XP_028700751.1 | 1069 | 142 | |||
XM_028844919.1 | XP_028700752.1 | 1033 | 142 | |||
XM_028844920.1 | XP_028700753.1 | 880 | 142 | |||
XM_028844921.1 | XP_028700754.1 | 942 | 142 | |||
XM_028844922.1 | XP_028700755.1 | 847 | 142 | |||
GenBank mRNA | JU336612.1 | - | 699 | 142 | ||
JU336613.1 | - | 873 | 142 | |||
JU336614.1 | - | 905 | 142 | |||
JU477425.1 | - | 935 | 142 | |||
JV044725.1 | - | 913 | 142 | |||
JV048386.1 | - | 925 | 142 | |||
JV630355.1 | - | 808 | 142 |

More Information
Set Membership: |
![]() ![]() |
