Assay Details
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257533.1 | NP_001244462.1 | 1005 | 112 | ||
XM_015133538.2 | XP_014989024.1 | 1190 | 112 | |||
XM_015133541.2 | XP_014989027.1 | 1063 | 112 | |||
XM_015133542.2 | XP_014989028.1 | 1019 | 112 | |||
XM_015133545.2 | XP_014989031.1 | 892 | 112 | |||
XM_015133547.2 | XP_014989033.1 | 793 | 112 | |||
XM_028844914.1 | XP_028700747.1 | 1343 | 112 | |||
XM_028844915.1 | XP_028700748.1 | 1260 | 112 | |||
XM_028844916.1 | XP_028700749.1 | 1218 | 112 | |||
XM_028844917.1 | XP_028700750.1 | 1106 | 112 | |||
XM_028844918.1 | XP_028700751.1 | 1169 | 112 | |||
XM_028844919.1 | XP_028700752.1 | 1133 | 112 | |||
XM_028844920.1 | XP_028700753.1 | 980 | 112 | |||
XM_028844921.1 | XP_028700754.1 | 1042 | 112 | |||
XM_028844922.1 | XP_028700755.1 | 947 | 112 | |||
GenBank mRNA | JU336612.1 | - | 799 | 112 | ||
JU336613.1 | - | 973 | 112 | |||
JU336614.1 | - | 1005 | 112 | |||
JU477425.1 | - | 1035 | 112 | |||
JV044725.1 | - | 1013 | 112 | |||
JV048386.1 | - | 1025 | 112 | |||
JV630355.1 | - | 908 | 112 |

More Information
Set Membership: |
![]() ![]() |
