Assay Details
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266438.1 | NP_001253367.1 | 458 | 116 | ||
XM_015143634.2 | XP_014999120.2 | 668 | 116 | |||
GenBank mRNA | JU322986.1 | - | 459 | 116 | ||
JU477203.1 | - | 458 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
