Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118163.2 | XP_014973649.2 | 1571 | 84 | ||
XM_015118164.2 | XP_014973650.1 | 997 | 84 | |||
XM_015118165.2 | XP_014973651.1 | 732 | 84 | |||
XM_015118166.2 | XP_014973652.1 | 1270 | 84 | |||
XM_015118167.2 | XP_014973653.2 | 596 | 84 | |||
XM_015118168.2 | XP_014973654.1 | 396 | 84 | |||
XM_028835841.1 | XP_028691674.1 | 1643 | 84 | |||
GenBank mRNA | JU319955.1 | - | 794 | 84 | ||
JU327976.1 | - | 635 | 84 | |||
JU334147.1 | - | 866 | 84 | |||
JU475277.1 | - | 661 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
