Assay Details
Gene Symbol: TNFSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266670.1 | NP_001253599.1 | 3 - 4 | 448 | 107 | |
XM_015116969.2 | XP_014972455.1 | 674 | 107 | |||
GenBank mRNA | JU477903.1 | - | 3 - 4 | 448 | 107 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
