Assay Details
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194644.2 | NP_001181573.1 | 278 | 66 | ||
XM_015118624.2 | XP_014974110.2 | 281 | 66 | |||
GenBank mRNA | JV052060.1 | - | 260 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
