Assay Details
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194644.2 | NP_001181573.1 | 5 - 6 | 804 | 66 | |
XM_015118624.2 | XP_014974110.2 | 807 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
