Assay Details
Gene Symbol: FYB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084072.4 | XP_001084072.2 | 1247 | 138 | ||
XM_001084428.4 | XP_001084428.1 | 1381 | 138 | |||
XM_015139942.2 | XP_014995428.1 | 1381 | 138 | |||
XM_015139943.2 | XP_014995429.1 | 1247 | 138 | |||
XM_028849414.1 | XP_028705247.1 | 1247 | 138 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
