Assay Details
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111442.4 | XP_001111442.1 | 867 | 133 | ||
GenBank mRNA | JU320778.1 | - | 516 | 133 | ||
JU473032.1 | - | 521 | 133 | |||
JV043521.1 | - | 520 | 133 |

More Information
Set Membership: |
![]() ![]() |
