Assay Details
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111442.4 | XP_001111442.1 | 940 | 101 | ||
XM_028845066.1 | XP_028700899.1 | 748 | 101 | |||
XM_028845067.1 | XP_028700900.1 | 788 | 101 | |||
GenBank mRNA | JU320778.1 | - | 5 - 6 | 589 | 101 | |
JU473032.1 | - | 5 - 6 | 594 | 101 | ||
JV043521.1 | - | 5 - 6 | 593 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
