Assay Details
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265987.1 | NP_001252916.1 | 3 - 4 | 716 | 117 | |
XM_028846630.1 | XP_028702463.1 | 875 | 117 | |||
XM_028846631.1 | XP_028702464.1 | 659 | 117 | |||
GenBank mRNA | JU322013.1 | - | 3 - 4 | 716 | 117 | |
JU478174.1 | - | 2 - 3 | 473 | 117 | ||
JV048616.1 | - | 3 - 4 | 716 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028846631.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
