Assay Details
Gene Symbol: ECI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125380.2 | XP_014980866.2 | 816 | 146 | ||
XM_015125381.2 | XP_014980867.2 | 1321 | 146 | |||
GenBank mRNA | JU325115.1 | - | 5 - 6 | 700 | 146 | |
JV045224.1 | - | 6 - 7 | 750 | 146 | ||
JV601734.1 | - | 6 - 7 | 762 | 146 | ||
JV667701.1 | - | 6 - 7 | 778 | 146 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
