Assay Details
Gene Symbol: HBEGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257469.1 | NP_001244398.1 | 477 | 111 | ||
GenBank mRNA | JU323062.1 | - | 477 | 111 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
