Assay Details
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130847.1 | XP_014986333.1 | 797 | 151 | ||
GenBank mRNA | JU334323.1 | - | 532 | 151 | ||
JU475425.1 | - | 473 | 151 | |||
JV046949.1 | - | 417 | 151 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
