Assay Details
Gene Symbol: KCNJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261696.1 | NP_001248625.1 | 2 - 3 | 1072 | 100 | |
GenBank mRNA | JU320252.1 | - | 2 - 3 | 1072 | 100 | |
JV601784.1 | - | 2 - 3 | 1119 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
