Assay Details
Gene Symbol: KCNJ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083031.3 | XP_001083031.1 | 605 | 99 | ||
XM_028845327.1 | XP_028701160.1 | 549 | 99 | |||
GenBank mRNA | JV683310.1 | - | 169 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001083031.3, XM_028845327.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
