Assay Details
Gene Symbol: LRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260541.1 | NP_001247470.1 | 1603 | 70 | ||
GenBank mRNA | JU326046.1 | - | 1584 | 70 | ||
JV044861.1 | - | 1603 | 70 | |||
JV616677.1 | - | 1727 | 70 | |||
JV616678.1 | - | 1727 | 70 | |||
JV667249.1 | - | 1621 | 70 |
Gene Symbol: SLC7A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV714996.1 | - | 1500 | 70 | ||
JV714997.1 | - | 1500 | 70 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
