Assay Details
Gene Symbol: MAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112833.2 | XP_014968319.1 | 1818 | 117 | ||
GenBank mRNA | JV545062.1 | - | 319 | 117 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
