Assay Details
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257605.2 | NP_001244534.1 | 3 - 4 | 374 | 132 | |
XM_028853315.1 | XP_028709148.1 | 475 | 132 | |||
XM_028853316.1 | XP_028709149.1 | 430 | 132 | |||
GenBank mRNA | JU323468.1 | - | 3 - 4 | 374 | 132 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
