Assay Details
Gene Symbol: PDE6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194606.1 | NP_001181535.1 | 3 - 4 | 455 | 149 | |
GenBank mRNA | JU334901.1 | - | 3 - 4 | 344 | 149 | |
JU472104.1 | - | 3 - 4 | 386 | 149 | ||
JV045081.1 | - | 3 - 4 | 356 | 149 | ||
JV547387.1 | - | 3 - 4 | 382 | 149 | ||
JV584037.1 | - | 3 - 4 | 349 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() |
