Assay Details
Gene Symbol: PRSS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266588.1 | NP_001253517.1 | 1 - 2 | 101 | 133 | |
GenBank mRNA | JU476439.1 | - | 1 - 2 | 101 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() |
