Assay Details
Gene Symbol: PRSS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266588.1 | NP_001253517.1 | 3 - 4 | 277 | 155 | |
XM_015125992.2 | XP_014981478.1 | 717 | 155 | |||
GenBank mRNA | JU476439.1 | - | 3 - 4 | 277 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() |
